Learn More
Water, UHPLC-MS
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 7732-18-5 | H2O | 18.015 g/mol
Supplier: Thermo Scientific W81
Description
Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, <-0.01 AU at 230nm, <-0.01 AU at 210nm, <-0.01 AU at 220nm, <-0.01 AU at 240nm, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, 2ppb max., 100 deg.C, 5ppb max., CHEBI:15377, Colorless, 1.000g/cm3, UHPLC-MS
Specifications
| ≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm | |
| 7 | |
| 7732-18-5 | |
| 100°C | |
| dihydrogen oxide, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| Clear Borosilicate Glass Bottle | |
| 17.5mmHg at 20°C | |
| 1 L |
| Water | |
| Colorless | |
| 0°C | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| UHPLC-MS | |
| ≤1ppm | |
| 1.000 g/cm3 | |
| Liquid |
RUO – Research Use Only