missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hughes Water Treatment Module
Hughes Water Treatment Module is for use with emergency tank showers
$5,889.56 - $7,813.42
Specifications
| Color | Yellow, Black |
|---|---|
| For Use With (Application) | Emergency Tank Showers |
| Includes | 1 Unit |
| Material | Ceramic Media |
Description
- These non-chemical disinfecting modules provide constant protection against infestations of pathogenic bacteria in water treatment systems.
- Immersed in water, the complex integral electrolytic activity of the ceramic media produces constant oxidizing, while the noble metal in the form of a fine matrix format produces ionization.
- The ionization of the noble metal is enhanced by the ceramic media and the combined effect is to act as a powerful disinfectant producing exceptional kill rates against pathogenic bacteria.
- The electrolytic activity of the ceramic media produces a chemical re-action that balances the pH of the water. This results in additional benefits in the form of scale inhibition and corrosion prevention.
- Optimum operating life of 3 years.
Compliance
Not ANSI Compliant
Warranty
1 Year
Specifications
| Yellow, Black | |
| Emergency Tank Showers | |
| 1 Unit | |
| Ceramic Media |