missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, TraceSELECT™, For Trace Analysis, Honeywell Riedel-de Haën™
TraceSELECT™, for trace analysis
Supplier: Honeywell Chemicals 953054X25L
Description
High purity solvents for speciation analysis by LC-ICP-MS
Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance
The blank values for metal traces in these solvents are in the ppb range or lower.
Specifications
| Water | |
| 7732-18-5 | |
| 0°C | |
| H2O | |
| NONH for all modes of transport | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| TraceSELECT | |
| Poly Bottle | |
| 4 x 2.5 L |
| 6 to 8 (at 20°C) | |
| Colorless | |
| 100°C | |
| MFCD00011332 | |
| 2050024 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| 18.02g/mol | |
| Odorless | |
| 1g/mL at 20°C | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| 18.015 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 962 | |
| oxidane |
| H2O | |
| MFCD00011332 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| CHEBI:15377 | |
| O |
Safety and Handling
RTECSNumber : 231-791-2