missing translation for 'onlineSavingsMsg'
Learn More

Water, TraceSELECT™, For Trace Analysis, Honeywell Riedel-de Haën™

TraceSELECT™, for trace analysis

Supplier:  Honeywell-Riedel-de Haen 953051L

 View more versions of this product

Catalog No. 60047765


Only null left
Explore available promotions

Description

Description

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

Specifications

Specifications

Water
7732-18-5
0°C
H2O
NONH for all modes of transport
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
O
18.015
CHEBI:15377
TraceSELECT
Glass Bottle
1 L
6 to 8 (at 20°C)
Colorless
100°C
MFCD00011332
2050024
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02g/mol
Odorless
1g/mL at 20°C
Liquid

Chemical Identifiers

7732-18-5
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane
H2O
MFCD00011332
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
CHEBI:15377
O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : 231-791-2

SDS
Documents

Documents

Promotions

Promotions