missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, for HPLC
CAS: 7732-18-5 | H2O | 18.015 g/mol
$165.11 - $165.11
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.015 |
| MDL Number | MFCD00011332 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| PubChem CID | 962 |
| ChEBI | CHEBI:15377 |
| IUPAC Name | oxidane |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389390025
|
Thermo Scientific Chemicals
389390025 |
2.5 L | Glass Bottle |
Each for $165.11
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCEcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Chemical Identifiers
| 7732-18-5 | |
| 18.015 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 962 | |
| oxidane |
| H2O | |
| MFCD00011332 | |
| dihydrogen oxide, dihydrogen monoxide | |
| CHEBI:15377 | |
| O |
Specifications
| 0.004 max. at 254nm, 0.009 max. at 210nm, 0.05 max. at 200nm | |
| Colorless | |
| 1μS/cm Max. | |
| H2O | |
| 15, 10237 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| 18.02 | |
| Glass Bottle | |
| 0.89 mPa.s (25°C) | |
| 2.5 L | |
| 1 | |
| Water |
| 7732-18-5 | |
| 100.0°C | |
| H2O | |
| MFCD00011332 | |
| dihydrogen oxide, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| HPLC | |
| 0.0003% max. | |
| 1.0000g/mL | |
| 1.3334 | |
| Liquid |
Safety and Handling
EINECSNumber : 231-791-2
RTECSNumber : ZC0110000
TSCA : TSCA
RUO – Research Use Only