missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Water, Sterile, Nuclease free, OmniPur™ Calbiochem™

Catalog No. 9801100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9801100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9801100ML Supplier MilliporeSigma™ Supplier No. 9801100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

DNase None detected
Grade Molecular Biology
Sterility Passes Test
Quantity 100 mL
Physical Form Liquid
Recommended Storage Ok to freeze