missing translation for 'onlineSavingsMsg'
Learn More

Water Sterile Filtered, Lab Chem

Catalog No. LC2671044
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC2671044 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC2671044 Supplier LabChem Supplier No. LC2671044
Only null left
  • Water Sterile Filtered Used in sterile environments

Chemical Identifiers

CAS 7732-18-5
Molecular Weight (g/mol) 18

Specifications

Percent Purity 100%
Grade Certified
Packaging Plastic Bottle
Quantity 4 x 4 L
Chemical Name or Material Water Sterile Filtered