missing translation for 'onlineSavingsMsg'
Learn More

Water, Standard, Apura™, MilliporeSigma™

Catalog No. M1092590250
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1092590250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1092590250 Supplier Technidata Supplier No. 1.09259.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Liquid water standard based on long-chain alcohol designed for daily titer control in Karl Fischer titration.

Chemical Identifiers

CAS 71-41-0

Specifications

Melting Point -78°C
Boiling Point 138°C (1013hPa)
Color Colorless
Density 0.82g/3 (20°C)
pH Neutral
Physical Form Liquid
Quantity 250 mL
Flash Point 49°C
Packaging Glass Bottle
Solubility Information 22g/L
Percent Purity 4.980 to 5.020 mg/mL
Grade Analytical
Chemical Name or Material Water in n-amyl alcohol
Show More Show Less
EINECSNumber 200-752-1