missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, RNase-free, DEPC Treated, Molecular Biology Grade, Ultrapure
CAS: 7732-18-5 | H2O | 18.015 g/mol
Supplier: Thermo Fisher Scientific J70783AP
Specifications
| Water | |
| 0°C | |
| H2O | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| Molecular Biology | |
| 1 | |
| 1.000 |
| 7732-18-5 | |
| 100°C | |
| MFCD00011332 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| 18.02 | |
| Nalgene Bottle | |
| 500 mL | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| 18.015 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 962 | |
| oxidane |
| H2O | |
| MFCD00011332 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| CHEBI:15377 | |
| O |
Safety and Handling
EINECSNumber : 231-791-2
RTECSNumber : ZC0110000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only