missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, RNase-free, DEPC Treated, Molecular Biology Grade, Ultrapure
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Formula Weight | 18.02 |
| Grade | Molecular Biology |
| Packaging | Nalgene Bottle |
| Density | 1 |
| Quantity | 1 L |
| Specific Gravity | 1.000 |
| Physical Form | Liquid |
| Chemical Name or Material | Water |
Safety and Handling
| EINECSNumber | 231-791-2 |
| RTECSNumber | ZC0110000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only