missing translation for 'onlineSavingsMsg'
Learn More

Water, Nuclease-free, Molecular Biology Grade, Ultrapure

Catalog No. aaj71786xcr
Change view
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
1 L
5 L
Packaging:
Nalgene Bottle
Upright carton
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ71786XCR 10 x 1 mL Nalgene Bottle
AAJ71786AE 100 mL Nalgene Bottle
AAJ71786AP 500 mL Nalgene Bottle
AAJ71786K2 1 L Nalgene Bottle
AAJ71786K8 5 L Upright carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAJ71786XCR Supplier Thermo Fisher Scientific Supplier No. J71786XCR
Only null left

CAS: 7732-18-5 | H2O | 18.015 g/mol

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Molecular Biology
Packaging Nalgene Bottle
Density 1
Quantity 10 x 1 mL
Specific Gravity 1.000
Physical Form Liquid
Chemical Name or Material Water
EINECSNumber 231-791-2
RTECSNumber ZC0110000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only