missing translation for 'onlineSavingsMsg'
Learn More

Water, Molecular Biology Grade, Sterile, Ricca Chemical

Catalog No. R91450001G
Change view
Click to view available options
Quantity:
1 L
Packaging:
Clear PET-G Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R91450001G 1 L Clear PET-G Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R91450001G Supplier Ricca Chemical Company Supplier No. R91450001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RNase Free, DNase Free, Protease Free, DEPC Free

  • Molecular biology grade water is free of Rnase, Dnase, Protease enzymes
  • DEPC free
  • Ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 5 to 7
Color Clear, Colorless
Melting Point 0°C
Boiling Point 100°C
DNase No
Solubility Information Miscible
Formula Weight 18.01
Percent Purity 100%
Grade Molecular Biology
Odor Odorless
Packaging Clear PET-G Bottle
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, Sterile, Dnase, Rnase, Protease, DEPC free
Show More Show Less
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .

NFPA

Health:0
Flammability:0
Instability:0