missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, MilliporeSigma
Description
LiChrosolv™ solvents exhibit high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them ideal for reproducible separations.
Specifications
Specifications
| Grade | UHPLC-MS |
| Packaging | Borosilicate Glass Bottle |
| Quantity | 1 L |
| Chemical Name or Material | Water |