missing translation for 'onlineSavingsMsg'
Learn More

Water, Low Organic, TraceGrade, Spectrum™ Chemical

Catalog No. 18614666
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614666 1 L Glass Bottle
18614667 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614666 Supplier Spectrum Chemical Mfg Cor Supplier No. W10041LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7732-18-5
Molecular Weight (g/mol) 18.02

Specifications

CAS Max % 1
Grade High Purity
Packaging Glass Bottle
Residue after Evaporation 1 ppm
Quantity 1 L
Chemical Name or Material Water, Low Organic

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.