missing translation for 'onlineSavingsMsg'
Learn More

Water, LC-MS Ultra CHROMASOLV™, Solstice

Catalog No. 60044839
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044839 1 L Glass Bottle
60044840 2 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044839 Supplier Solstice Supplier No. 142631L

CHROMASOLV™ LC-MS Ultra, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 6 to 8 (at 20°C)
Color Colorless
Melting Point 0°C
Boiling Point 100°C
UN Number NONH for all modes of transport
Beilstein 2050024
Formula Weight 18.02g/mol
Grade CHROMASOLV™ LC-MS Ultra
Packaging Glass Bottle
Density 1.000g/mL at 3.98°C
Quantity 1 L
Refractive Index n20/D 1.34(lit.)
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
EINECSNumber 231-791-2
RTECSNumber ZC0110000