missing translation for 'onlineSavingsMsg'
Learn More

Water, LC-MS Grade

Catalog No. p-7052285
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47146K2 1 L
AA47146K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA47146K2 Supplier Thermo Scientific Chemicals Supplier No. 047146K2
Only null left

Mobile phase solvent in LC-MS analysis | CAS: 7732-18-5 | H2O | 18.015 g/mol

It is used as mobile phase solvent in LC-MS analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Beilstein 2050024
Merck Index 14,10039
Solubility Information Soluble in water. Insoluble in polar liquids.
Formula Weight 18.02
Grade LC-MS
Odor Odorless
Density 1.000 g/mL
Quantity 1 L
Refractive Index 1.34
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
EINECSNumber 231-791-2
RTECSNumber ZC0110000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only