missing translation for 'onlineSavingsMsg'
Learn More

Water for Injection, EP/USP, Sterile Grade, Cytiva Hyclone™

Catalog No. p-7161867
Change view
Click to view available options
Quantity:
20 L
4 x 4 L
200 L
Packaging:
Bioprocess Bag
HDPE Jug
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
Z0104 4 x 4 L HDPE Jug
WFIEPZ20L 20 L Bioprocess Bag
WFIEPZ2XL 200 L Bioprocess Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Z0104 Supplier Cytiva Supplier No. Z0104
Only null left

Water for Injection, EP/USP, is a terminally distilled, sterile, non-pyrogenic preparation which contains no bacteriostat, antimicrobial agent or added buffer.

  • Complies with the USP and EP monographs for water for injection packaged in bulk for commercial use
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications
  • Widely used in applications where a high quality sterile water is required

For Manufacturing or Research Use only, not intended for Diagnostic or Therapeutic Use.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear, Colorless
Endotoxin <0.25 EU/mL
Grade EP/USP
Packaging HDPE Jug
Sterility Sterile
Certificate of Analysis Yes
Quantity 4 x 4 L
ShelfLife 36 Months
Recommended Storage 2-30°C