missing translation for 'onlineSavingsMsg'
Learn More

Water, HPLC, LabChem

Catalog No. LC267604
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC267604 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC267604 Supplier LabChem Supplier No. LC267604

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HPLC Water is ideal for use in high-performance liquid chromatography, separations, and mobile phase chromatography.

  • For Use in High-Performance Liquid Chromatography, Separations, Mobile Phase.
  • 0.2 Micron Filtered.
  • LC Suitable Absorbance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Water
IUPAC Name water
SMILES O

Specifications

Absorbance 0.01 AU/cm (Maximum at 200 nm)
pH 7
Color Clear, Colorless
Melting Point 0°C
Boiling Point 100°C
CAS Max % 100%
Conductivity 1.5 uS/cm (Conductivity at 25C Maximum)
Percent Purity 100%
Grade HPLC
Packaging Glass Bottle
Residue after Evaporation 1 ppm (Maximum)
Sterility Non-sterile
Filtered Through 0.2 Micron Filtered
Density 1.00 mg/mL
Certificate of Analysis Yes
Quantity 4 L
Chemical Name or Material Water
Show More Show Less
ShelfLife 24 Months
Recommended Storage Room Temperature