missing translation for 'onlineSavingsMsg'
Learn More

Water, HPLC, J.T. Baker™

Catalog No. 02004558
Change view
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004558 1 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02004558 Supplier Avantor Supplier No. 421802

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High purity solvents for use in analytical and preparative separations

    For use in Liquid Chromatography (HPLC & UHPLC) & Spectrophotometry; For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Formula Weight 18
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1 L
Specific Gravity 1L = 1.00kg
Physical Form Liquid
Chemical Name or Material Water