missing translation for 'onlineSavingsMsg'
Learn More

Water Hardness Titrating Solution, Reagents

Water Hardness Titrating Solution 1 mL = 1 mg CaCO3, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$92.96 - $679.79

Specifications

Color Colorless
For Use With (Application) Water Analysis
Sterility Non-sterile
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-333-387
View Documents
Labchem Inc
CW00090010F
10 L
Each for $410.00
Only null left
 
01-333-386
View Documents
Labchem Inc
CW0009001A
1 L
Each for $92.96
Only null left
 
01-333-385
View Documents
Labchem Inc
CW00090020F
20 L
Each for $679.79
Only null left
 
01-333-384
View Documents
Labchem Inc
CW0009004A
4 L
Each for $159.79
Only null left
 
Description

Description

Specifications

Specifications

Colorless
Water Analysis
Non-sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'