Learn More
Water Hardness Titrating Solution, Reagents
Water Hardness Titrating Solution 1 mL = 1 mg CaCO3, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CW0009001A
Specifications
| Colorless | |
| Water Analysis | |
| 1 L |
| 1 L | |
| Poly Bottle | |
| Non-sterile |