missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Hardness Buffer, without Magnesium EDTA, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Water Hardness Buffer |
| Name Note | Without Magnesium EDTA |
| CAS | 7732-18-5, 1336-21-6, 12125-02-9 |
| CAS Min % | 62.0, 31.88, 4.19 |
| Color | Colorless |
| CAS Max % | 64.0, 33.18, 4.36 |
| Concentration | 1X |
| For Use With (Application) | Water Analysis |
| Physical Form | Liquid |
| Quantity | 1 L |
| Show More |