missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water Hardness Buffer, Certified, LabChem™
Supplier: LabChem LC268002
Specifications
| Water Hardness Buffer | |
| 64,30,5.5,0.5 | |
| Nitrogen oxides; Carbon monoxide; Carbon dioxide | |
| 1 L | |
| C2H7NO | |
| Poly Bottle | |
| 61.08 |
| Yellow | |
| 141-43-5,7647-01-0,14402-88-1 | |
| Liquid | |
| Passes Test | |
| Odorless | |
| Soluble in water | |
| Certified |