missing translation for 'onlineSavingsMsg'
Learn More

Water Hardness Buffer, Certified, LabChem™

Catalog No. LC268001
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC268001 500 mL
LC268002 1 L
LC268004 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC268001 Supplier LabChem Supplier No. LC268001
Only null left

Specifications

Chemical Name or Material Water Hardness Buffer
Color Yellow
CAS Max % 64, 30, 5.5, 0.5
CAS 141-43-5, 7647-01-0, 14402-88-1
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula C2H7NO
Odor Odorless
Packaging Poly Bottle
Solubility Information Soluble in water
Formula Weight 61.08
Grade Certified
Show More Show Less