missing translation for 'onlineSavingsMsg'
Learn More

Water, For LC-MS, OmniSolv™, MilliporeSigma™

Catalog No. p-7082939
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MWX00016 1 L Glass Bottle
MWX00011 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MWX00016 Supplier Technidata Supplier No. WX00016
Only null left

Recommended Storage

Store at room temperature

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Boiling Point 100°C (1013 hPa)
Formula Weight 18.02
Packaging Glass Bottle
Vapor Pressure 23 hPa (20 °C)
Density 1.00 g/3 (20 °C)
Quantity 1 L
Physical Form Liquid
Recommended Storage Store at room temperature