missing translation for 'onlineSavingsMsg'
Learn More

Water, For LC-MS, OmniSolv™, MilliporeSigma™

Catalog No. mwx00011
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
MWX00011 4 L Glass Bottle
MWX00016 1 L Glass Bottle
2 options

Catalog No. MWX00011

Supplier: Technidata WX00011

Only null left

Recommended Storage

Store at room temperature

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Boiling Point 100°C (1013 hPa)
Formula Weight 8.02
Packaging Glass Bottle
Vapor Pressure 23 hPa (20°C)
Quantity 4 L
Physical Form Liquid
RTECSNumber ZC0110000
Recommended Storage Store at room temperature