missing translation for 'onlineSavingsMsg'
Learn More

Water, For HPLC Gradient Analysis, Spectrophotometry and Gas Chromatography, OmniSolv™, MilliporeSigma™

Catalog No. MWX0004P1
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glas Bottle (Pre packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MWX0004P1 4 L Glas Bottle (Pre packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MWX0004P1 Supplier Technidata Supplier No. WX0004P1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH Neutral (at 20°C)
Melting Point 0°C
Boiling Point 100°C (1013 hPa)
Formula Weight 18.02
Grade Meets Reagent Specifications for testing USP/NF monographs
Odor None
Packaging Glas Bottle (Pre packed)
Residue after Evaporation 1ppm
Vapor Pressure 23 hPa (20°C)
Viscosity 0.952 mPa.s at 20°C
Density 1.00 g/cc at 20°C
Quantity 4 L
Physical Form Liquid
For Use With (Application) Chromatography
Chemical Name or Material Water
Show More Show Less
RTECSNumber ZC0110000
storageNote1 0 - 13 Other liquids and solids
Recommended Storage Store at room temperature