missing translation for 'onlineSavingsMsg'
Learn More

Water (Environmental Grade), Fisher Chemical™

Catalog No. W114
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
W114 4 L Amber Glass
W111 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. W114 Supplier Seastar Chemicals Inc Supplier No. S010901TFIA43
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Packaged in precleaned amber glass bottles to protect product integrity

  • With less than 20 ppb TOC, Fisher Chemical recommends this product for the most stringent total organic carbon requirements
  • Tested for gradient suitability, optical absorbance and conductivity makes this product suitable for all environmental testing

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.01 AU Maximum (200nm), 0.005 AU Maximum (254nm), 0.005 AU Maximum (300nm), 0.005 AU Maximum (400nm)
pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Linear Formula H2O
Formula Weight 18.02
Grade Environmental
Packaging Amber Glass
Residue after Evaporation 1 ppm Maximum
Vapor Pressure 17.5mmHg at 20°C
Density 1.000g/cm³
Certificate of Analysis Certificate of Lot Analysis included with each bottle.
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .

NFPA

Health:0
Flammability:0
Instability:0