missing translation for 'onlineSavingsMsg'
Learn More

Water, (DNASE, RNASE free), Fisher BioReagents™

Catalog No. p-23724
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Nalgene Poly Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP248450 50 mL Nalgene Poly Bottle
BP2484100 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP248450 Supplier Fisher BioReagents Supplier No. BP248450
Only null left

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 100mL, Vapor Pressure: 17.5mmHg at 20 deg.C, 100 deg.C, CHEBI:15377, Gray, Colorless, 1.000g/cm3, DNase free, Filtered through a 0.2-micron filter., 18.02, Low hazard for usual industrial or commercial handling.

Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
DNase DNase free
Merck Index 15, 10237
Formula Weight 18.02
Packaging Nalgene Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.2-micron filter.
Treatment(s) DEPC-treated and autoclaved.
Density 1.000g/cm³
Quantity 50 mL
Physical Form Liquid
Chemical Name or Material Water, DNASE, RNASE free
Show More Show Less
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult