missing translation for 'onlineSavingsMsg'
Learn More

Water, Distilled, Reagent Grade, Ricca Chemical

Catalog No. p-3252621
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Cubitainer
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
918032 1 L Amber Glass Bottle
91801 4 L Amber Glass Bottle
91805 20 L Cubitainer
R918000055E 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 918032 Supplier Ricca Chemical Company Supplier No. 918032
Only null left

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Color Colorless
Conductivity max. 1.5 μS/cm
Synonym dihydrogen oxide, dihydrogen monoxide
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Percent Purity 100%
Grade Reagent
Packaging Amber Glass Bottle
Sterility Non-sterile
Quantity 1 L
Chemical Name or Material Water, Distilled
Show More Show Less
ShelfLife 18 months