missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Water, DEPC Treated, Sterile, Nuclease-Free, OmniPur™, Calbiochem™,

Catalog No. 9601100ML
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
9601100ML 100 mL Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9601100ML Supplier MilliporeSigma™ Supplier No. 9601100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for RNA applications (Prepared with DEPC)

Recommended Storage

+15°C to +30°C.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Grade Molecular Biology
Packaging Plastic Bottle
Quantity 100 mL
Physical Form Liquid
Recommended Storage +15°C to +30°C.