missing translation for 'onlineSavingsMsg'
Learn More

Water, deionized MilliporeSigma™

Catalog No. 23749999
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
23749999 20 L Cubitainer
23749998 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23749999 Supplier Technidata Supplier No. US16442/88

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water; deionized; distilled; Type II; HARLECO; ; H2O

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Packaging Cubitainer
Quantity 20 L
Chemical Name or Material Water