missing translation for 'onlineSavingsMsg'
Learn More

Water, Deionized, ASTM Type I, Certified ACS Grade, LabChem™

Catalog No. LC267402
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
Packaging:
Cubitainer
Poly Bottle
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC267402 1 L Poly Bottle
LC267404 4 L Poly Bottle
LC267406 10 L Cubitainer
LC267405 20 L Cubitainer
LC26740200 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC267402 Supplier LabChem Supplier No. LC267402

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
CAS Max % 100
Conductivity ≤ 0.056uS/cm
Linear Formula H2O
Solubility Information Soluble in water
Formula Weight 18.02
Grade Certified ACS
Packaging Poly Bottle
Viscosity 1.004cSt
Density 0.99823g/mL
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, Deionized, ASTM Type I
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
EINECSNumber 231-791-2
Recommended Storage Room Temperature