missing translation for 'onlineSavingsMsg'
Learn More

Water ChromAR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MK67951
View Documents
Avantor
AVA679506
1L Glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.015
dihydrogen oxide, dihydrogen monoxide
CHEBI:15377
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
oxidane

Specifications

7732-18-5
dihydrogen oxide, dihydrogen monoxide
O
18.015
CHEBI:15377
Glass
Water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
oxidane
962
18.02
1L
Videos
SDS
Documents

Documents