missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva
Eliminate endotoxin concerns with HyClone™ Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.
Supplier: Cytiva SH30529LS
Description
- Endotoxin-free; contains no detectable gram-negative bacteria
- Meets < 0.005 EU/mL endotoxin by turbidimetric assay
Specifications
| 6 x 1 L |
Safety and Handling
ShelfLife : 24 months
Recommended Storage : 2° to 30°C