missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva

Catalog No. p-7058039
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
6 x 500 mL
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3052901 100 mL
SH3052902 500 mL
SH3052903 1 L
SH30529FS 6 x 500 mL
SH30529LS 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SH3052901 Supplier Cytiva Supplier No. SH30529.01
Only null left

Eliminate endotoxin concerns with HyClone™ Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.

  • Endotoxin-free; contains no detectable gram-negative bacteria
  • Meets < 0.005 EU/mL endotoxin by turbidimetric assay

Specifications

pH 5.0 to 7.0
Quantity 100 mL