missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva

Eliminate endotoxin concerns with HyClone™ Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.

Supplier:  Cytiva SH30529FS

 View more versions of this product

Catalog No. SH30529FS

Only null left
Explore available promotions

Description

Description

  • Endotoxin-free; contains no detectable gram-negative bacteria
  • Meets < 0.005 EU/mL endotoxin by turbidimetric assay
Specifications

Specifications

5.0 to 7.0
<0.005EU/mL
Liquid
Water, Cell Culture Grade (Endotoxin-Free)
6 x 500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 30°C

SDS
Documents

Documents

Promotions

Promotions