missing translation for 'onlineSavingsMsg'
Learn More

Water (Biotech Grade), Fisher BioReagents

Catalog No. p-23703
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly CUBE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP24854 4 L Poly CUBE
BP248520 20 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP24854 Supplier Fisher BioReagents Supplier No. BP24854
Only null left
Biotech-grade water can be used for several biological test procedures.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Merck Index 15, 10237
Formula Weight 18.02
Grade Biotech
Packaging Poly CUBE
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.2-micron filter.
Density 1.000g/cm³
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult