missing translation for 'onlineSavingsMsg'
Learn More

Water, for HPLC and Spectrophotometry, Solstice

Catalog No. 6003135
Change view
Click to view available options
Quantity:
20 L
Packaging:
Returnable Nowpak™ Ii
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6003135 20 L Returnable Nowpak™ Ii
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6003135 Supplier Solstice Supplier No. NS36520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Boiling Point 100°C
CAS Max % 100.0000%
Beilstein 2050024
Formula Weight 18
Grade HPLC
Packaging Returnable Nowpak™ Ii
Density 0.9971 g/cm3
Quantity 20 L
Chemical Name or Material Water, B&J Brand™, for HPLC and Spectrophotometry
ShelfLife 547 days from date of manufacture