missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, ACS reagent
CAS: 7732-18-5 | H2O | 18.02 g/mol
$99.40 - $99.40
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC434400010
|
Thermo Scientific Chemicals
434400010 |
1 L | Plastic Bottle |
Each for $99.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7732-18-5 | |
| H2O | |
| 18.02 | |
| dihydrogen oxide, dihydrogen monoxide |
Specifications
| Passes Test, 0.01 max. at 210nm, 0.005 max. at 254nm, 0.01 max. at 200nm, 0.005 max. at 300nm | |
| 7732-18-5 | |
| 2μS/cm Max. | |
| H2O | |
| dihydrogen oxide, dihydrogen monoxide | |
| 18.02 | |
| Plastic Bottle | |
| 1.0000g/mL | |
| 1.3334 | |
| Crystalline Powder |
| White | |
| 100°C | |
| H2O | |
| 15,10237 | |
| 18.02 | |
| ACS Reagent | |
| 0.89 mPa.s (25°C) | |
| 1 L | |
| 1 | |
| Water, For analysis ACS |
Safety and Handling
EINECSNumber : 231-791-2
RUO – Research Use Only