missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, ACS Reagent Grade, ASTM Type I, ASTM Type II, Ricca Chemical
Specifications
Specifications
| Percent Purity | 100% |
| Grade | ACS |
| Packaging | Amber Glass Bottle |
| Quantity | 4 L |
| Chemical Name or Material | Water, ASTM Type I, ASTM Type II |