missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, ACS Reagent Grade, ASTM Type I, ASTM Type II, Ricca Chemical
Specifications
Specifications
| pH | <1 |
| Color | Colorless |
| Melting Point | ∼0°C |
| CAS Min % | 100 |
| Boiling Point | ∼100°C |
| CAS Max % | 100 |
| Solubility Information | Miscible in water |
| Formula Weight | 18.02 |
| Percent Purity | 100% |
| Grade | ACS |
| Show More |
Safety and Handling
| ShelfLife | 18 months |
| Recommended Storage | Ambient |