missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% Trifluoroacetic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™

Catalog No. p-3850220
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LS119500 500 mL Glass Bottle
LS1191 1 L Wide Neck Amber Bottle
LS119212 2.5 L Wide Neck Amber Bottle
LS1194 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LS119500 Supplier Thermo Fisher Scientific Supplier No. LS119 500
Only null left
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • TFA is a common mobile phase additive that can function as an ion pair reagent

Chemical Identifiers

CAS 7732-18-5 , 76-05-1
PubChem CID 962
ChEBI CHEBI:15377

Specifications

Absorbance Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm
Assay Percent Range 99.0 to 103.0 %
Molecular Formula C2HF3O2
MDL Number MFCD00004169
Merck Index 15, 9847
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES OC(=O)C(F)(F)F
IUPAC Name trifluoroacetic acid
Molecular Weight (g/mol) 114.02
Percent Purity 0.095 to 0.105% (v/v) Trifluoroacetic Acid
Grade OPTIMA™ LC-MS
Packaging Glass Bottle
Residue after Evaporation ≤1ppm
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material 0.1% Trifluoroacetic Acid in Water (v/v), Solvent Blends
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0