Learn More
Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade
Supplier: Fisher Chemical LS118212
Description
- Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
- Pre-mixed for ease-of-use
- Low metal ion content to ensure minimal metal contamination
- Formic acid (FA), a common mobile phase additive that provides protons for LC/MS analysis
- FA is also known to improve the peak shapes of the resulting separation
Specifications
| 0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm | |
| 2 | |
| 7732-18-5,64-18-6 | |
| ≥99.0 % | |
| 3297 | |
| dihydrogen oxide, dihydrogen monoxide | |
| O | |
| 18.015 | |
| CHEBI:15377 | |
| Wide Neck Amber Bottle | |
| 1g/cm³ | |
| Liquid |
| 0.1% Formic Acid in Water (v/v) | |
| Solvent Blends | |
| 100°C | |
| H2O | |
| 15, 4269 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| oxidane | |
| 962 | |
| OPTIMA™ LC-MS | |
| 1ppm max. | |
| 2.5 L |
Chemical Identifiers
| 7732-18-5 | |
| 18.015 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 962 | |
| oxidane |
| H2O | |
| 3297 | |
| dihydrogen oxide, dihydrogen monoxide | |
| CHEBI:15377 | |
| O |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0