missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade

Catalog No. LS118500
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LS118500 500 mL Glass Bottle
LS1181 1 L Wide Neck Amber Bottle
LS118212 2.5 L Wide Neck Amber Bottle
LS1184 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LS118500 Supplier Thermo Fisher Scientific Supplier No. LS118 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • Formic acid (FA); common mobile phase additive that provides protons for LC/MS analysis
  • FA is also known to improve the peak shapes of resulting separation

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 3297
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
pH 2
Boiling Point 100°C
Assay Percent Range ≥99.0 %
Merck Index 15, 4269
Grade OPTIMA™ LC-MS
Packaging Glass Bottle
Residue after Evaporation 1ppm max.
Density 1g/cm³
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material 0.1% Formic Acid in Water (v/v), Solvent Blends
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0