missing translation for 'onlineSavingsMsg'
Learn More

Mobile phase for HPLC, Water with formic acid (0.1%), HPLC Grade, Fisher Chemical

Catalog No. HB5234
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
HB5234 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HB5234 Supplier Thermo Fisher Scientific Supplier No. HB5234
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Ideal mobile phase blend developed for use with HPLC instruments
  • Pre-mixed for ease-of-use
  • Formic acid (FA) is a common mobile phase additive used in HPLC separations
  • FA controls the pH of the mobile phase by functioning as a buffer
  • FA is also known to improve the peak shapes of the resulting separation

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 2.7
Boiling Point 100°C
Assay Percent Range ≥98.5 %
Grade HPLC
Packaging Glass Bottle
Density 1g/cm³
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water with 0.1% Formic Acid (v/v)
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0