missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% Formic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC, Honeywell Burdick & Jackson

Catalog No. p-7112038
Change view
Click to view available options
Quantity:
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Glass Bottle, Gl45
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6003079 6 x 1 L Glass Bottle, Gl45
6003080 4 x 2.5 L Glass Bottle, Gl45
6003081 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6003079 Supplier Honeywell Inc Supplier No. LC4521
Only null left

Chemical Identifiers

CAS 7732-18-5 , 64-18-6 , 7732-18-5
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

CAS Max % 99.9000%, 0.1000%
Formula Weight 46.03
Grade HPLC
Density 0.997 g/cm3
Quantity 6 x 1 L
Packaging Glass Bottle, Gl45
Chemical Name or Material Water with 0.1% Formic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC
ShelfLife 365 days from date of manufacture