missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with 0.1% Formic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC, Honeywell Burdick & Jackson
Supplier: Honeywell-Burdick and Jackson LC45225
Spécifications
| Water with 0.1% Formic Acid | |
| 7732-18-5,64-18-6,7732-18-5 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| HPLC | |
| 4 x 2.5 L |
| LabReady™ Solvent Blend, for LC-MS and HPLC | |
| 99.9000%,0.1000% | |
| 46.03 | |
| 0.997 g/cm3 | |
| Glass Bottle, Gl45 |
Identifiants chimiques
| 7732-18-5 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
Sécurité et manipulation
missing translation for 'shelfLife' : 365 days from date of manufacture