missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Buffer Components (1 and 2)

Wash Buffer Components (1 and 2) are an individual component of the ViewRNA™ ISH Tissue Assay Kits, 1-Plex.

Supplier:  Invitrogen™ QVT0502

Catalog No. QVT0502


Only null left

Description

Description

Wash Buffer Components (1 and 2) are an individual component of the ViewRNA™ ISH Tissue Assay Kits, 1-Plex (Cat. No. QVT0050). It is an aqueous buffered solution containing detergent, and can be used for 24 assays (108 mL). Store at -15°C to -30°C.

Specifications

Specifications

Wash Buffer Components (1 and 2), 108 mL

Sufficient for 24 assays.
Store at -15°C to -30°C.
Aqueous buffered solution
Wash Buffer Component
24 Assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.