Learn More
Fisherbrand™ Walk-In Fume Hood Work Floor
Fisherbrand™ Walk-In Fume Hood Work Floor is designed to be used with Fisherbrand Walk-in Fume Hoods.
 View more versions of this product
Catalog No. 06000618
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • 1.25 in. thick modified black epoxy resin work floors are an option for all Walk-In Fume Hoods
  • Designed to retain spills.
Specifications
Chemical Identifiers
Epoxy
Fisherbrand Walk-In Fume Hoods
3.2 cm
Black
1.25 in.
Specifications
Epoxy
37 in.
Fisherbrand Walk-In Fume Hoods
3.2 cm
152.4 cm
Black
93.98 cm
1.25 in.
60 in.
Product Suggestions
Product Suggestions